Axygen™ Microplate Sealing Film and Tapes

Offer great economy for disposable ease

Manufacturer: Axygen Scientific BF444

 View more versions of this product

Catalog No. 14-223-301

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »


    Specifications

    Material Rayon
    No. per Case 1000
    No. per Pack 100.0
    Product Type Breathable Sealing Film
    Length (Metric) 153mm
    Length (English) 6.02 in.
    Thickness 50μm
    Width (English) 3.24 in.
    Width (Metric) 82.3mm
    Dimensions (L x W) 6.02 x 3.24 in. (153 x 82.3mm)
    Applications Designed for applications ranging from PCR and real-time PCR to ELISA and cell culture, allow effective gas exchange for cellular and bacterial cultivation, while preventing contamination
    Sterile No
    Temperature Range -20° to 80°C

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit