Learn More
Axygen™ Microplate Sealing Film and Tapes
Offer great economy for disposable ease.
Supplier: Axygen™ HSFAS300L
Description
For applications ranging from PCR and Real-Time PCR to ELISA and cell culture, Axygen films are an easy and cost-effective way to seal plates and enhance automation. Used to seal multi-well microplates.
Aluminum
- Provides exceptional seal on multi-well plates with varied temperatures and reagents
- With compression mats, films and tapes seal during thermocycling
- Ideal for light-sensitive samples and its uniformly applied adhesive provides optimum well sealing
- Pierceable, PCR-AS-200
Specifications
Aluminum | |
4.52 in. | |
1/Cs. | |
4.52 x 196 in. (115 x 500 m) | |
500 m | |
Agilent/V11 | |
-80°C to +104°C |
Non-sterile | |
115 mm | |
1/Pk. | |
196 in. | |
Designed for applications ranging from PCR and real-time PCR to storage, ideal for light-sensitive samples | |
Sealing Film Roll |