Axygen™ Microtube Racks: 80-Well Axyracks
Ideal for storage and handling of PCR/Micro/Screw cap tubes
Manufacturer: Axygen Scientific R80OF
 View all options for this product
Catalog No. 14-223-359
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Color Fluorescent Orange
    Holds 1.5 to 2mL Tubes
    Material Polypropylene
    Positions 80.0
    Quantity 25/case
    Product Type Microtube rack
    • Holds eighty 1.5/2.0mL micro and screw cap tubes
    • Polypropylene; resistant to most lab chemicals and solvents
    • For cryogenic storage of tubes
    • Conical bottom wells for superior stability of conical tubes
    • Each well numbered for ease of sample identification
    • Frosted side writing panel
    • Available in a variety of fluorescent and non-fluorescent colors
    • Packaging: 25 Racks/Cs.
  • Description & Specifications
Related Products
Product Content Feedback