Axygen™ Multi Well High Profile Reagent Reservoirs Available on GSA/VA Contract for Federal Government customers only.

Manufactured with virgin polypropylene for excellent chemical resistance

Manufacturer: Axygen Scientific RESMW8HP

 View more versions of this product

Catalog No. 14-222-407

  Check Availability
Add to Cart

    Specifications

    Product Type Reagent Reservoir
    Includes 8-channel troughs
    Volume (English) Well 0.33 oz.
    Material Polypropylene
    Certifications/Compliance ANSI/SLAS, RNase/DNase-free, non-pyrogenic
    Packaging 25/Cs.
    Capacity 2.7 oz., 80mL
    Color Clear
    Disposable Yes
    Volume (Metric) Well 10mL
    Sterile No
    Capacity (English) 2.7 oz.
    Capacity (Metric) 80mL

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit