Axygen™ Multi Well Low Profile Reagent Reservoirs Available on GSA/VA Contract for Federal Government customers only.

Manufactured with virgin polypropylene for excellent chemical resistance

Manufacturer: Axygen Scientific RESMW12LPSI

 View more versions of this product

Catalog No. 14-222-405

  Check Availability
Add to Cart

    Specifications

    Certifications/Compliance ANSI/SLAS, RNase/DNase-free, non-pyrogenic
    Capacity 0.71 oz., 21mL
    Color Clear
    Volume (English) Well 0.059 oz.
    Capacity (Metric) 21mL
    Material Polypropylene
    Packaging Individually Wrapped; 25/Cs.
    Disposable Yes
    Product Type Reagent Reservoir
    Includes 12-channel troughs
    Volume (Metric) Well 1.75mL
    Sterile Yes
    Capacity (English) 0.71 oz.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit