Axygen™ Multi Well Low Profile Reagent Reservoirs Available on GSA/VA Contract for Federal Government customers only.

Manufactured with virgin polypropylene for excellent chemical resistance

Manufacturer: Axygen Scientific RESMW8LP

 View more versions of this product

Catalog No. 14-222-409

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Specifications

    Material Polypropylene
    Certifications/Compliance ANSI/SLAS, RNase/DNase-free, non-pyrogenic
    Packaging 25/Cs.
    Capacity 1.08 oz., 32mL
    Color Clear
    Disposable Yes
    Product Type Reagent Reservoir
    Includes 8-channel troughs
    Volume (Metric) Well 4mL
    Volume (English) Well 0.135 oz.
    Sterile No
    Capacity (English) 1.08 oz.
    Capacity (Metric) 32mL

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit