Axygen™ Multimek Robotic Tips: 200µL

Fits Multimek 'Type II' or CCS-style head

Manufacturer: Axygen Scientific MT200R

 View more versions of this product

Catalog No. 14-222-193

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    Product Type Robotic Tip
    Material Polypropylene
    Quantity 96 Tips/Rack
    For Use With (Equipment) Beckman™ Coulter™ MultiMek™ automated system
    Sterility Nonsterile
    Tip Style Racked
    Autoclavable Yes
    Packaging Rack
    Certifications/Compliance Non-ANSI
    For Use With (Application) Unique diamond polished molding technology use no mold-releasing agents, minimizes wastage of precious samples and reagents while handling, nucleic acid, nuclease and endotoxin-safe for complete protection against contamination
    Volume (Metric) 200μL
    Color Clear
    Rack Color Blue

    Robotic tip's unique diamond-polished molding technology uses no mold-releasing agents. Minimizes wastage of precious samples and reagents while handling. Nucleic acid, nuclease and endotoxin-safe for complete protection against contamination.

    • Tip volume: 200µL
    • Tip color: Clear
    • Rack color: Red and blue

    Order Info

    Racked: 96 tips/rack, 10 racks/pack, 5 packs/case

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit