Learn More
Axygen™ Multimek Type II Robotic Tips
Robbins-style tips for Beckman Multimek 'Type II'
Manufacturer:  Axygen Scientific RMT200CR
 View more versions of this product
Catalog No. 14-222-958
  Check Availability
Add to cart
Specifications
Description
  • Presterilized tips are exposed to e-beam radiation for complete security without degrading the plastic or performance
  • Volume range: 200µL
  • Available in racked and racked sterile formats
  • Packaging: 96 tips/rack, 10 racks/pack, 5 packs/case
Description & Specifications
Specifications
Robotic Tip
Nonsterile
Racked
96 Tips/Rack
Exposed to e-beam radiation for complete security without degrading the plastic or performance
200μL
Clear
Rack
RNase, DNase, and endotoxin free
Beckman™ Coulter™ Multimek™ II Systems