Axygen™ PCR Tubes with 0.2mL Dome Cap

Thin and consistent wall thickness for precise thermal transfer

Manufacturer: Axygen Scientific PCR02DB

 View more versions of this product

Catalog No. 14-222-272

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    Volume (Metric) 0.2mL
    Format Tube
    For Use With (Equipment) Standard 96-well blocks of real-time PCR instruments and thermal transfer applications
    Capacity (Metric) 0.2mL
    Type Thin Wall, Domed Cap PCR Tubes
    Quantity 1000/Pk., 10Pk/Cs.
    Sterile No
    Closure Material Polypropylene
    Cap Type Domed
    Shape Conical
    Certifications/Compliance RNase, DNase, pyrogen-free
    Color Blue
    Material Polypropylene

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit