Axygen™ PCR Tubes with 0.2mL Dome Cap Available on GSA/VA Contract for Federal Government customers only.

Thin and consistent wall thickness for precise thermal transfer

Manufacturer: Axygen Scientific PCR-02D-SP

 View all options for this product

Catalog No. 14-222-278


This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Type Thin Wall, Domed Cap PCR Tubes
    Cap Type Domed
    Certifications/Compliance RNase, DNase, pyrogen-free
    Color Spectrum
    Material Polypropylene
    Volume (Metric) 0.2mL
    Quantity 1000/Pk., 10Pk/Cs.
    Sterile No
    Closure Material Polypropylene
    Format Tube
    For Use With (Equipment) Standard 96-well blocks of real-time PCR instruments and thermal transfer applications
    Shape Conical
    Capacity (Metric) 0.2mL

    Quantitative PCR compatible tubes for a wide range of Real-Time PCR instruments.

    • PCR tubes with 0.2mL volume
    • Tubes with caps sealed full-length; insertion cap ensures tight fit, eliminates sample evaporation
    • Choose from a wide variety of colors

  • Description & Specifications

Related Products

Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit