Axygen™ PCR Tubes with 0.5mL Flat Cap Available on GSA/VA Contract for Federal Government customers only.

Thin and consistent wall thickness for precise thermal transfer

Manufacturer: Axygen Scientific PCR05O

 View more versions of this product

Catalog No. 14-222-295

  Check Availability
Add to Cart

    Specifications

    Type Thin Wall PCR Tube
    Volume (Metric) 0.5mL
    Quantity 1000 Pack
    Sterile No
    Closure Material Polypropylene
    Cap Type Flat
    Format Tube
    For Use With (Equipment) Standard 96-well blocks of real-time PCR instruments and thermal transfer applications
    Shape Conical
    Capacity (Metric) 0.5mL
    Certifications/Compliance RNase, DNase, pyrogen-free
    Color Orange
    Material Polypropylene

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit