Learn More
Axygen™ PCR Tubes with 0.5mL Flat Cap Available on GSA/VA Contract for Federal Government customers only.
Thin and consistent wall thickness for precise thermal transfer
Manufacturer:  Axygen Scientific PCR05Y
 View more versions of this product
Catalog No. 14-222-299
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Description
Quantitative PCR compatible tubes for a wide range of Real-Time PCR instruments.
  • Tubes with caps sealed full-length; insertion cap ensures tight fit, eliminates sample evaporation
  • Choose from a wide variety of colors
  • Certified RNase-/DNase-free
  • Nonpyrogenic
  • Nonsterile
  • Manufactured from polypropylene
Description & Specifications
Specifications
Thin Wall PCR Tube
RNase, DNase, pyrogen-free
Polypropylene
1000/Pk., 10Pk./Cs.
Polypropylene
Standard 96-well blocks of real-time PCR instruments and thermal transfer applications
0.5mL
Flat
Yellow
0.5mL
No
Tube
Conical