Learn More
Axygen™ PerkinElmer™/Packard Robotic Tips: Maxymum Recovery
Ideal for MiniTrak, PlateTrak and Evolution P-3
Manufacturer:  Axygen Scientific PK50LR
 View more versions of this product
Catalog No. 14-222-369
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications
Description
Robotic tips uses no mold-releasing agents. Nucleic acid, nuclease and endotoxin-safe for complete protection against contamination.
  • Tip volume: 20 to 200µL
  • Tip color: clear
  • Rack color: green
Description & Specifications
Specifications
Robotic Tip
Nonsterile
Green
Round
Rack
RNase, DNase-free, ANSI
PerkinElmer™ Packard™ Janus™/Evolution P3™/MiniTrak™/PlateTrak™ automated liquid handling systems and MDT-96 P235, MDT-96 P50 and MDT-384 30μL head
50μL
Clear
Racked
Polypropylene
96 Tips/Rack
Maxymum Recovery™, non-pyrogenic, used for stringent specifications under strict process controls, nucleic acid, nuclease and endotoxin-safe for complete protection against contamination