Axygen™ Black Automation Tips for PerkinElmer™, 96-Well
Ideal for Janus™ MiniTrak, PlateTrak™ and Evolution P3. Axygen™ Black Automation Tips for PerkinElmer™, 96-Well are racks of non-sterile, non-filtered, 200µL pipette tips. They are constructed of black polypropylene for light-sensitive samples.
Manufacturer: Axygen™ PK200BKR
Catalog No. 14-223-201
  Check Availability
Add to Cart
    Specifications
    For Use With (Equipment) PerkinElmer™ Packard™ Janus™/Evolution P3™/MiniTrak™/PlateTrak™ automated liquid handling systems and MDT-96 P235, MDT-96 P50 and MDT-384 30μL head
    Product Type For Packard's Plate Track
    Material Polypropylene
    Quantity 96 Tips/Rack
    Filtered No
    Sterility Nonsterile
    Shape Round
    Packaging Rack
    Certifications/Compliance RNase, DNase-free
    For Use With (Application) Non-pyrogenic, used for stringent specifications under strict process controls, nucleic acid, nuclease and endotoxin-safe for complete protection against contamination
    Volume (Metric) 200μL
    Color Black
    Rack Color Green
    • Robotic tips use no mold-releasing agents
    • Nucleic acid, nuclease and endotoxin-safe for complete protection against contamination
    • Tips are certified RNase-/DNase-free and nonpyrogenic
    • Assembled via automation under strict process controls to ensure consistent performance and quality
    • Compatible with PerkinElmer™ MDT-96 P235 head for:
      • Janus™
      • Evolution P3 
      • Minitrak
      • PlateTrak™
  • Description & Specifications
Related Products
Product Content Feedback