Axygen™ Automation Tips for PerkinElmer™, 96-Well Available on GSA/VA Contract for Federal Government customers only.
Ideal for Janus™ MiniTrak, PlateTrak™ and Evolution P3. Axygen™ Automation Tips for PerkinElmer™, 96-Well are constructed of clear polypropylene. Available in sterile/non-sterile, filtered/non-filtered and wide bore options, from 20 to 200µL.
Manufacturer: Axygen™ PKF180RS
 View more versions of this product
Catalog No. 14-222-374
  Check Availability
Add to Cart
    Specifications
    For Use With (Equipment) PerkinElmer™ Packard™ Janus™/Evolution P3™/MiniTrak™/PlateTrak™ automated liquid handling systems and MDT-96 P235, MDT-96 P50 and MDT-384 30μL head
    Product Type Filter, Racked, Presterilized
    Material Polypropylene
    Quantity 96 Tips/Rack
    Sterility Sterile
    Shape Round
    Packaging Rack
    Certifications/Compliance RNase, DNase-free, ANSI
    For Use With (Application) Non-pyrogenic, used for stringent specifications under strict process controls, nucleic acid, nuclease and endotoxin-safe for complete protection against contamination
    Volume (Metric) 180μL
    Color Clear
    Rack Color Green
    Assembled via automation under strict process controls to ensure consistent performance and quality
    • Robotic tips use no mold-releasing agents
    • Nucleic acid, nuclease and endotoxin-safe for complete protection against contamination
    • Tips are certified RNase-/DNase-free and nonpyrogenic
    • Compatible with PerkinElmer™ MDT-96 P235, MDT-96 P50 and MDT-384 30µL head for:
      • Janus™
      • Evolution P3
      • Minitrak
      • PlateTrak™
    Multiple options
    • Presterilized and non-sterile tips
    • With or without aerosol-resistant polyethylene filters
    • Wide bore series for working with viscous materials, genomic DNA or delicate cell cultures
    • Available in a range of volumes: 20, 30, 50, 180 and 200µL
  • Description & Specifications
Related Products
Product Content Feedback