Learn More
Axygen™ Universal Fit Micropipette Tips
Axygen™ Universal Fit Pipette Tips are compatible with most research-grade single- and multi-channel micropipettes, and are certified to be DNase-/RNase-free and nonpyrogenic.
$369.00
Specifications
Clean Claims Dnase-, Rnase-, Pyrogen-, Human gDNA-, PCR-Inhibition-Free
Autoclavable Autoclavable
Filtered Non-filtered
Volume (Metric) 200 μL
For Use With (Equipment) Most brands of single and multichannel pipettes
View More Specs
Products 1
Catalog Number Mfr. No. Color Sterility Tip Style Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Color Sterility Tip Style Packaging Price Quantity & Availability  
14-222-720
View Documents
Axygen™
T200CSTKS
Clear Non-sterile Beveled Stack Rack Case of 4800 for $369.00
Add to cart
Description
Description
  • RNase-/DNase free
  • Nonpyrogenic
  • Human gDNA- and PCR-inhibition-free
  • Available in a variety of sizes
  • Available with a variety of tip styles including beveled, fine point, and wide bore
  • Available sterile/non-sterile
  • Available in bulk, racked, or stack rack packaging
  • Available with Maxymum Recovery™ surface for improved accuracy and maximum sample recovery
Specifications
Specifications
Dnase-, Rnase-, Pyrogen-, Human gDNA-, PCR-Inhibition-Free
Non-filtered
Most brands of single and multichannel pipettes
Autoclavable
200 μL
PP (Polypropylene)