Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axygen™ Power Supply Adapters for 7cm Gel Box

Accessory for use with 7cm horizontal gel box

Manufacturer:  Axygen Scientific HGB7AD

 View more versions of this product

Catalog No. 14-223-980


Add to cart

Specifications

Specifications

4mm to 4mm power supply adapter
7cm gel box
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit