Axygen™ Presterilized Conical Centrifuge Tubes Available on GSA/VA Contract for Federal Government customers only.

Graduated tubes with conical bottom

Manufacturer: Axygen Scientific SCT15ML25S

 View more versions of this product

Catalog No. 14-222-960

  Check Availability
Add to Cart

    Specifications

    Product Type Centrifuge Tube
    Capacity (Metric) 15mL
    Color Clear
    Material Polypropylene
    Max. Force 12000xG
    Sterile Yes
    For Use With (Application) For sample identification
    Cap Style Screw Cap
    Includes Screw cap
    Packaging Bag
    Shape Conical
    Autoclavable No

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit