Learn More
Axygen™ Robotic Pipette Tips Available on GSA/VA Contract for Federal Government customers only.
Compatible with Hamilton MICROLAB™ STAR and Nimbus workstations
Manufacturer:  Axygen Scientific HT50CBKHTRS
 View more versions of this product
Catalog No. 14-223-705
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Automation Tip
Sterile
Yes
Round
Hanging Tip Rack, 24 x 96
DNase, RNase, pyrogen free
Hamilton™ Microlab™ star, starlet, starplus and Nimbus™ workstations
50μL
Black
Non-filtered
Polypropylene
96/rack, 24 racks/cs.
The conductive, liquid level sensing tips are available in four sizes and specially designed for use with Hamilton™ liquid handling platforms, they enable precision pipetting across a wide range of applications