Learn More
Axygen™ Robotic Pipette Tips
Compatible with Hamilton MICROLAB™ STAR and Nimbus workstations
Manufacturer:  Axygen™ HT1000CBKHTR
 View more versions of this product
Catalog No. 14-223-710
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
The conductive, liquid level sensing tips are available in four sizes and specially designed for use with Hamilton liquid handling platforms. They enable precision pipetting across a wide range of applications.
  • Sizes: 10, 50, 300 and 1000µL
  • Sterile or nonsterile; filter tip also available
  • Packaging options include hanging tip rack, traditional deck-seating rack or nested/stacked format
  • Fully field automated; manufactured under strict process control
  • 100% automated assembly to ensure consistent performance and quality
  • Hanging Tip Rack: fits Hamilton STAR, STARlet™ and STARplus™
  • Deck-seating Rack: for MICROLAB STAR Series and Hamilton Nimbus
Specifications
Specifications
Hamilton™ MICROLAB™ STAR, Hamilton™ MICROLAB™ STARlet, Hamilton™ MICROLAB™ STARplus, Hamilton™ MICROLAB™ Nimbus
Automation
Non-Filtered
Black
Round
DNase, RNase, and Pyrogen free
Hamilton™ MICROLAB™ STAR, Hamilton™ MICROLAB™ STARlet, Hamilton™ MICROLAB™ STARplus, Hamilton™ MICROLAB™ Nimbus
Hanging Tip Rack
1000μL
Non-sterile
Yes
PP (Polypropylene)
The conductive, liquid level sensing tips are available in four sizes and specially designed for use with Hamilton™ liquid handling platforms, they enable precision pipetting across a wide range of applications