Axygen™ Robotic Pipette Tips Available on GSA/VA Contract for Federal Government customers only.

Compatible with Hamilton MICROLAB™ STAR and Nimbus workstations

Manufacturer: Axygen Scientific HT10CBKHTR

 View more versions of this product

Catalog No. 14-223-701

  Check Availability
Add to Cart

    Specifications

    Product Type Automation Tip
    Material Polypropylene
    Packaging Hanging Tip Rack, 24 x 96
    Quantity 96/rack, 24 racks/cs.
    For Use With (Equipment) Hamilton™ Microlab™ star, starlet, starplus and Nimbus™ workstations
    Sterility Nonsterile
    Autoclavable Yes
    Tip Style Non-filtered
    Shape Round
    Certifications/Compliance DNase, RNase, pyrogen free
    For Use With (Application) The conductive, liquid level sensing tips are available in four sizes and specially designed for use with Hamilton™ liquid handling platforms, they enable precision pipetting across a wide range of applications
    Volume (Metric) 10μL
    Color Black

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit