Axygen™ Screw Caps with O-rings Available on GSA/VA Contract for Federal Government customers only.

Select looped or non-looped format

Manufacturer: Axygen Scientific SCOLPB

 View more versions of this product

Catalog No. 14-222-490

  Check Availability
Add to Cart

    Specifications

    Color Blue
    Sterile No
    For Use With (Equipment) Both skirted and non-skirted variations of the 0.5mL, 1.5mL, and 2mL microcentrifuge tubes
    Format Looped
    Includes O-ring
    Material Polypropylene

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit