Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axygen™ Single Well High Profile Reagent Reservoirs

Manufactured with virgin polypropylene for excellent chemical resistance

Manufacturer:  Axygen Scientific RESSW384HP

 View more versions of this product

Catalog No. 14-222-417

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Description

Translucent polypropylene reservoirs are designed with a standard SBS footprint for compatability with robotic systems.

Diamond Shaped-well Series: 96- or 384- collection pools at the bottom surface help minimize dead volume

Description & Specifications

Specifications

No
282mL
ANSI/SLAS, RNase/DNase-free, non-pyrogenic
9.53 oz., 282mL
Yes
384-bottom troughs
9.53 oz.
Polypropylene
25/Cs.
Clear
Reagent Reservoir
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit