Learn More
Axygen™ MaxyClear Snaplock Microtubes, 0.60mL Available on GSA/VA Contract for Federal Government customers only.
Ensure an efficient seal that stays closed during centrifugation. Axygen™ MaxyClear Snaplock Microtubes, 0.60mL provide excellent visibility and feature a piercing port for syringe access. Available in a wide variety of colors. Maxymum Recovery™ and sterile options available.
Manufacturer:  Axygen™ MCT060B
 View more versions of this product
Catalog No. 14-222-142
  Check Availability
Add to cart
Specifications
Description
  • MaxyClear polypropylene provides excellent visibility and is available in a variety of colors
  • Snaplock cap assures efficient seal that stays closed during centrifugation or boiling
  • 0.28mm piercing port in the center of the cap allows easy access to sample by syringe needle
  • Frosted cap surfaces for labeling and frosted panels on the side for writing
  • Can withstand up 14,000 RCF
  • Boil-proof and fully autoclavable
  • Available in a variety of options to meet the needs of different applications:
    • Maxymum Recovery™ option provides excellent accuracy and prevents wastage of precious samples
    • Clear, sterile tubes available for applications requiring sterility
    • Available in a range of colors or assorted packs to support color-coding
Description & Specifications
Specifications
Snap Cap
Polypropylene
0.6mL
Polypropylene
No
0.1mL
RNase, DNase, pyrogen-free
Yes
Blue
14000xG
0.1 to 0.6mL
1000 Tubes/Pk., 10 Packs/Cs.