Axygen™ Speciality Tips Available on GSA/VA Contract for Federal Government customers only.

Suitable for Axypet-16 channel pipettor for 384/1536-well formats, Multi-channel Plus and macrovolume

Manufacturer: Axygen Scientific T384LR

 View more versions of this product

Catalog No. 14-222-752

  Check Availability
Add to Cart

    Specifications

    Product Type Maxymum Recovery™ Tip
    Material Polypropylene
    For Use With (Equipment) 16-channel Pipettor
    Sterility Nonsterile
    Autoclavable Yes
    Packaging Rack
    Certifications/Compliance RNase, DNase, and endotoxin free
    Volume (Metric) 0.5 to 60μL
    Color Clear

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit