Learn More
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
Axygen™ Storage Microplates
Used for sample collection, in vitro growth chambers and long-term storage.
Manufacturer:  Axygen Scientific P96450VW
 View more versions of this product
Catalog No. 14-222-244
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Description
Choose from a variety of configurations:
  • 48 deep-well rectangular plate with pyramid bottom
  • 96 deep-well plate with square wells
  • 384 deep-well diamond plate, with square wells
  • 96-well assay plate with round bottom (SBS footprint) or V-bottom (SBS footprint)
  • 24 deep-well rectangular plate
Description & Specifications
Specifications
Yes
White
Polypropylene
96
-112°F
96-Well Storage V-bottom Microplate
ANSI, SLAS 1-2004
Sample collection, in-vitro growth chambers and long term storage
4000xG
No
-80°C
550μL