Axygen™ Storage Microplates Available on GSA/VA Contract for Federal Government customers only.

Manufacturer: Axygen Scientific PDW10ML24CS

 View more versions of this product

Catalog No. 14-222-350

  Check Availability
Add to Cart

    Specifications

    For Use With (Application) Sample collection, in-vitro growth chambers and long term storage
    Material Polypropylene,Polypropylene
    No. of Wells 24
    Temperature Range (English) -112°F
    Temperature Range (Metric) -80°C
    Type 24 Rectangular Deep-Well Storage Microplate
    Volume (Metric) Well 10000μL,10000μL
    Well Design Rectangular
    Color Clear
    Autoclavable Yes
    Max. RCF 4000xG
    Sterile Yes
    Well Shape Rectangular

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit