Axygen™ Storage Microplates Available on GSA/VA Contract for Federal Government customers only.

Manufacturer: Axygen Scientific P96450RY

 View more versions of this product

Catalog No. 14-222-238

  Check Availability
Add to Cart

    Specifications

    Autoclavable Yes
    For Use With (Application) Sample collection, in-vitro growth chambers and long term storage
    Material Polypropylene,Polypropylene
    Max. RCF 4000xG
    No. of Wells 96
    Sterile No
    Temperature Range (English) -112°F
    Temperature Range (Metric) -80°C
    Type 96-Well Round-bottom Storage Microplate
    Certifications/Compliance ANSI, SLAS 1-2004
    Volume (Metric) Well 550μL,550μL
    Well Design Round
    Color Yellow
    Well Shape Round

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit