Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Axygen™ Tecan™ Evo MCA96 Compatible Nested Pipet Tips
768 tips for use with Tecan Evo MCA96 multi-channel arm
Supplier: Axygen™ EV50NTR
Description
- Tip format: 96 tip
- Tip volume range: 50 μL to 200 μL
- Tip material: Clear polypropylene
- RNase-/DNase-/human gDNA-free
- Nonpyrogenic
- Available options:
- With or without aerosol-resistant filters
- Nonsterile or sterile - Compatible heads: MCA96
- Rack format: ANSI/SLAS
- Rack material: Polypropylene
Specifications
50 μL | |
Orange | |
Nested tip rack | |
Tecan EVO MCA96 | |
Polypropylene |
Non-sterile | |
Round | |
40 Racks/Cs. | |
Tecan |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction