Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axygen™ Tube Closures

Designed to fit standard 10 to 16mm glass and plastic test tubes

Manufacturer:  Axygen Scientific ACPEB

 View more versions of this product

Catalog No. 14-222-000

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Specifications

Polyethylene
Test Tube Cap
10, 12, 13, 16mm tubes
10 to 16mm
Blue
No
0.39 to 0.63 in.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit