Learn More
Axygen™ Wellpro™ Robotic Tips Available on GSA/VA Contract for Federal Government customers only.
Clear tips
Manufacturer:  Axygen Scientific TWP200R
 View more versions of this product
Catalog No. 14-222-862
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Robotic Tip
Nonsterile
Gray
Round
Rack
RNase, DNase-free, Non-ANSI
ProGroup WellPro™ 3000-96/3000 automated liquid handling system with 8/12-channel head
200μL
Clear
Racked
Polypropylene
96 Tips/Rack
Non-pyrogenic, used for stringent specifications under strict process controls