Axygen™ Zymark™ Rapid Plate/Sciclone and Tecan™ TEMO Acquarius Genmate: 100µL
Manufacturer: Axygen Scientific ZT100LRS
 View more versions of this product
Catalog No. 14-222-906
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    Product Type Robotic Tip
    Material Polypropylene
    Quantity 96 Tips/Rack
    For Use With (Equipment) Tecan™ Freedom Evo™, TeMo and Aquarius with MCA96 head, Zymark™/Caliper RapidPlate™/SciClone™ with HV96 head/CAS-4200-1ch.-20-150μL/sec. head
    Sterility Sterile
    Autoclavable Yes
    Tip Style Racked
    Shape Round
    Packaging Rack
    Certifications/Compliance Non-ANSI
    Volume (Metric) 100μL
    Color Clear
    Rack Color Gray
  • Description & Specifications
Related Products
Product Content Feedback