Learn More
Axygen™ Zymark™ Rapid Plate/Sciclone and Tecan™ TEMO Acquarius Genmate: 100µL Available on GSA/VA Contract for Federal Government customers only.
Manufacturer:  Axygen Scientific ZT100LR
 View more versions of this product
Catalog No. 14-222-905
  Check Availability
Add to cart
Specifications
Description
Packaging: 96 Tips/Rack, 10 Racks/Pack, 5 Packs/Case
Description & Specifications
Specifications
Robotic Tip
Nonsterile
Gray
Racked
Polypropylene
96 Tips/Rack
Tecan™ Freedom Evo™, TeMo and Aquarius with MCA96 head, Zymark™/Caliper RapidPlate™/SciClone™ with HV96 head/CAS-4200-1ch.-20-150μL/sec. head
100μL
Clear
Yes
Round
Rack
Non-ANSI