Axygen™ Zymark™ Rapid Plate/Sciclone and Tecan™ TEMO Acquarius Genmate: 170µL
Manufacturer: Axygen Scientific ZTF205WBRS
 View more versions of this product
Catalog No. 14-222-971
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    Product Type Filtered Robotic Tip
    Volume (Metric) 200μL
    Sterility Sterile
    Color Clear
    Rack Color Blue
    Autoclavable Yes
    Tip Style Filter, Racked
    Shape Wide Bore
    Material Polypropylene
    Packaging Rack
    Quantity 96 Tips/Rack
    Certifications/Compliance Non-ANSI
    For Use With (Equipment) Tecan™ Freedom Evo™, TeMo and Aquarius with MCA96 head, Zymark™/Caliper RapidPlate™/SciClone™ with HV96 head/CAS-4200-1ch.-20-150μL/sec. head
    • Clear tips
    • Light Green, Blue and Gray racks
  • Description & Specifications
Related Products
Product Content Feedback