Learn More
Axygen™ Zymark™ Rapid Plate/Sciclone and Tecan™ TEMO Acquarius Genmate: 200µL
Manufacturer:  Axygen Scientific ZT200LR
 View more versions of this product
Catalog No. 14-222-909
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Description
  • Color-coded racks
  • 96 Tips/Rack, 10 Racks/Pack, 5 Packs/Case
Description & Specifications
Specifications
Robotic Tip
Nonsterile
Blue
Racked
Polypropylene
96 Tips/Rack
Maxymum Recovery™ to further improve accuracy and precision while reducing sample loss
200μL
Clear
Yes
Round
Rack
Non-ANSI
Tecan™ Freedom Evo™, TeMo and Aquarius with MCA96 head, Zymark™/Caliper RapidPlate™/SciClone™ with HV96 head/CAS-4200-1ch.-20-150μL/sec. head