Learn More
Tocris Bioscience AZ 12216052
Positive allosteric modulator at mGlu8
Manufacturer: Tocris Bioscience 4832/10
Description
Positive allosteric modulator at mGlu8 receptors. Exhibits anxiolytic effects in mouse models of anxiety.This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
2-[[(4-Bromophenyl)methyl]thio]-N-[4-(1-methylpropyl)phenyl]acetamide | |
C19H22BrNOS | |
10mg | |
0.97 |
1290628-31-7 | |
392.35 | |
Target: Group III mGluR Modulators |
Safety and Handling
Recommended Storage : Store at -20°C
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.