Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience AZ 12216052
Positive allosteric modulator at mGlu8
Manufacturer: Tocris Bioscience 4832/50
This item is not returnable.
View return policy
Description
Positive allosteric modulator at mGlu8 receptors. Exhibits anxiolytic effects in mouse models of anxiety.Specifications
2-[[(4-Bromophenyl)methyl]thio]-N-[4-(1-methylpropyl)phenyl]acetamide | |
C19H22BrNOS | |
50mg | |
0.97 |
1290628-31-7 | |
392.35 | |
Target: Group III mGluR Modulators |
Safety and Handling
Recommended Storage : Store at -20°C
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title