Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Azadibenzocyclooctyne-NHS ester, with 4 carbon linker

Click to view available options
Quantity:
25mg
100mg
Applications
Azadibenzocyclooctyne-NHS ester, with 4 carbon linker is used in various applications like protein-peptide conjugates, antibody-enzyme or antibody-drug conjugates, protein or peptide-oligonucleotide conjugates and surface modification. It is used to introduce cyclooctyne moiety in to biomolecules and amine containing compounds.

Solubility
Soluble in dimethyl formamide, terahydrofuran, dimethyl sulfocide and dichloromethane.

Notes
Moisture and heat sensitive. Store in cool place. Incompatible with strong oxidizng agents.
Quantity 100mg
Color White to Gray
Sensitivity Moisture Sensitive
Solubility Information Soluble in DMF, DMSO, THF and DCM
Formula Weight 402.4
Physical Form Solid
Chemical Name or Material Azadibenzocyclooctyne-NHS ester, with 4 carbon linker

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

storageNote1 : -30° to -10°C