Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Azadibenzocyclooctyne-PEG12 biotin conjugate

Click to view available options
Quantity:
10mg
25mg
C55H83N5O16S
ADIBO-PEG12-biotin conjugate
C55H83N5O16S
ADIBO-PEG12-biotin conjugate
  • An ADIBO-biotin conjugate with a very hydrophilic PEG12 spacer
  • The PEG12 spacer eliminates aggregation and precipitation when labeling antibodies and other biological materials
Color White to Gray
Quantity 25mg
Formula Weight 1102.34
Physical Form Solid
Chemical Name or Material Azadibenzocyclooctyne-PEG12 biotin conjugate

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Recommended Storage : −30° to −10°C