Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Azadibenzocyclooctyne-PEG4-alcohol

Click to view available options
Quantity:
25mg
100mg
C29H36N2O7
MFCD22380739
ADIBO-PEG4-alcohol
C29H36N2O7
MFCD22380739
ADIBO-PEG4-alcohol

ADIBO-containing reagent with PEG4 spacer and alcohol terminus

Color Yellow
Quantity 100mg
Formula Weight 524.61
Physical Form Solid or Viscous Liquid
Chemical Name or Material Azadibenzocyclooctyne-PEG4-alcohol

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Recommended Storage : −30° to −10°C