Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Azadibenzocyclooctyne-PEG4-NHS ester

Click to view available options
Quantity:
10mg
25mg
100mg
MFCD22380747
MFCD22380747
Applications
Azadibenzocyclooctyne-PEG4-NHS ester is applied for protein-peptide conjugation, peptide-small molecule conjugation, 18F radiolabeling, protein-oligonucleotide conjugation and surface modification.

Solubility
Soluble in DCM, DMF, DMSO and THF.

Notes
Moisture sensitive. Incompatible with oxidizing agents and heat.
Color Orange to Brown
Quantity 25mg
Sensitivity Moisture Sensitive
Formula Weight 649.69
Physical Form Oil
Chemical Name or Material Azadibenzocyclooctyne-PEG4-NHS ester

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

storageNote1 : -30° to -10°C