Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Azaperone, Acros Organics™

Click to view available options
Quantity:
1g
5g
25g
1649-18-9
C19H22FN30
327.4
XTKDAFGWCDAMPY-UHFFFAOYSA-N
azaperone,stresnil,fluoperidol,suicalm,azaperon,azeperone,eucalmyl,sedaperone vet,azaperona,azaperonum
15443
1-(4-fluorophenyl)-4-(4-pyridin-2-ylpiperazin-1-yl)butan-1-one
C1CN(CCN1CCCC(=O)C2=CC=C(C=C2)F)C3=CC=CC=N3
1649-18-9
C19H22FN30
327.4
XTKDAFGWCDAMPY-UHFFFAOYSA-N
azaperone,stresnil,fluoperidol,suicalm,azaperon,azeperone,eucalmyl,sedaperone vet,azaperona,azaperonum
15443
1-(4-fluorophenyl)-4-(4-pyridin-2-ylpiperazin-1-yl)butan-1-one
C1CN(CCN1CCCC(=O)C2=CC=C(C=C2)F)C3=CC=CC=N3
Quantity 25g
Infrared Spectrum Authentic
Loss on Drying 0.5% max. (60°C, 4 h) (vacuum)
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in methanol, chloroform
Formula Weight 327.4
Chemical Name or Material Azaperone

GHS H Statement
Toxic if swallowed.

GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

GHS Signal Word: Danger

EINECSNumber : 216-715-8