Learn More
Cayman Chemical Azaserine, 115-02-6, 10 mg
SDP
Supplier:  Cayman Chemical 1483410
Encompass_Preferred
A tumor-inhibiting antibiotic that inhibits the glutamine-dependent amidotransferases involved in nucleotide biosynthesis, N-formylglycineamidine ribotide synthetase and glucosamine-6-phosphate isomerase; also inhibits the hexosamine biosynthetic pathway, which has been shown to protect against hyperglycemic endothelial damage
Catalog No. NC1415416
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.