Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Azathioprine

Click to view available options
Quantity:
1g
5g
10g
446-86-6
C9H7N7O2S
277.262
MFCD00069203
LMEKQMALGUDUQG-UHFFFAOYSA-N
azathioprine, azothioprine, imuran, azathioprin, azatioprin, azamun, azanin, imurel, ccucol, imurek
2265
CHEBI:2948
6-(3-methyl-5-nitroimidazol-4-yl)sulfanyl-7H-purine
CN1C=NC(=C1SC2=NC=NC3=C2NC=N3)[N+](=O)[O-]
446-86-6
C9H7N7O2S
277.262
MFCD00069203
LMEKQMALGUDUQG-UHFFFAOYSA-N
azathioprine, azothioprine, imuran, azathioprin, azatioprin, azamun, azanin, imurel, ccucol, imurek
2265
CHEBI:2948
6-(3-methyl-5-nitroimidazol-4-yl)sulfanyl-7H-purine
CN1C=NC(=C1SC2=NC=NC3=C2NC=N3)[N+](=O)[O-]
Solubility
Soluble in Dichloromethane and dimethyl sulfoxide.

Notes
Store at -20°C. Incompatible with oxidizing agents. Protect from heat.
Color Yellow
Quantity 1g
Merck Index 14,902
Solubility Information Soluble in Dichloromethane and dimethyl sulfoxide.
Formula Weight 277.26
Physical Form Crystalline Powder
Chemical Name or Material Azathioprine

GHS H Statement
H350-H302-H315-H319-H335
May cause cancer.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P201-P202-P261-P264b-P270-P271-P280i-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P332+P313-P362-P501c

H302-H315-H319-H335-H350

EINECSNumber : 207-175-4

RTECSNumber : UO8925000

TSCA : No

Recommended Storage : Store at -20°C