Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Azathioprine, USP, 98-102%, Spectrum™

Click to view available options
Quantity:
1 g
5 g
25 g
100 g
1 kg
10 kg
Packaging:
Amber Glass Bottle
Poly Pail
446-86-6
C9H7N7O2S
277.26
LMEKQMALGUDUQG-UHFFFAOYSA-N
6-[(1-methyl-4-nitro-1H-imidazol-5-yl)sulfanyl]-7H-purine
CN1C=NC(=C1SC1=C2NC=NC2=NC=N1)[N+]([O-])=O
446-86-6
C9H7N7O2S
277.26
LMEKQMALGUDUQG-UHFFFAOYSA-N
6-[(1-methyl-4-nitro-1H-imidazol-5-yl)sulfanyl]-7H-purine
CN1C=NC(=C1SC1=C2NC=NC2=NC=N1)[N+]([O-])=O

A1383, 446-86-6, C9H7N7O2S

Spectrum™ Azathioprine, USP is an immunosuppressive drug used in autoimmune diseases and organ transplants. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Amber Glass Bottle
Ignition Residue 0.001
Loss on Drying 0.01
Quantity 1 g
CAS Max % 1
Percent Purity 98 to 102%