Learn More
Azzota Corp AZD-5363 DFS Item
Manufacturer:  Azzota Corp C1058
AZD-5363 is an orally available inhibitor of the serine/threonine protein kinase AKT (protein kinase B) with potential antineoplastic activity. Please request if need more quantities
Catalog No. 50-161-3065