Learn More
Azzota Corp AZD-8055 DFS Item
Manufacturer:  Azzota Corp C1065
AZD-8055 inhibits the serine/threonine kinase activity of mTOR, resulting in decreased expression of mRNAs necessary for cell cycle progression, which may induce cell cycle arrest and tumor cell apoptosis. Please request if need more quantities
Catalog No. 50-161-3069