Learn More
Azzota Corp AZD-8330 DFS Item
Manufacturer:  Azzota Corp C1067
AZD-8330 specifically inhibits mitogen-activated protein kinase kinase 1 (MEK or MAP/ERK kinase1), resulting in inhibition of growth factor-mediated cell signaling and tumor cell proliferation. Please request if need more quantities
Catalog No. 50-161-3071