Learn More
Selleck Chemical LLCSupplier Diversity Partner AZD8330 10mg 869357-68-6 ARRY704  DFS Item
Manufacturer:  Selleck Chemical LLCSupplier Diversity Partner S213410MG
AZD8330 (ARRY704) is a novel, selective, non-ATP competitive MEK 1/2 inhibitor with IC50 of 7 nM. Phase 1.
Catalog No. 50-714-9