Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Azelaic Acid, 85%, Spectrum™

Click to view available options
Quantity:
25 g
100 g
1 kg
5 kg
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
123-99-9
C9H16O4
188.22
BDJRBEYXGGNYIS-UHFFFAOYSA-N
nonanedioic acid
OC(=O)CCCCCCCC(O)=O
123-99-9
C9H16O4
188.22
BDJRBEYXGGNYIS-UHFFFAOYSA-N
nonanedioic acid
OC(=O)CCCCCCCC(O)=O

A2215, 123-99-9, C9H16O4

Spectrum™ Azelaic Acid , found in rye, wheat and barley, and is an organic compound. It is found in many hair and skin related products. It is also used in lubrication and plasticizers as well as in the medical world to treat acne and is categorized as a dicarboxylic acid.
CAS Max % 100%
Percent Purity 85%
Packaging Amber Glass Bottle
Quantity 100 g